Dithallium selenide ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Thallium(I) selenide
36296.04
36296.09
ALFA36296.04EA
221
SGD
ALFA36296.04
ALFA36296.09
Dithallium selenide ≥99.9% (metals basis)
Dithallium selenide
Purity: 99.9%
CAS No.: 15572-25-5
Molecular Formula: Tl2Se
Formula Weight: 487.70
Physical Form: 10mm & Down Lump
Density: 9.05
Melting Point: ca 340°
Solubility: Insoluble in water, acids
Merck Reference: 14,9267
UN No.: 1707
MDL No.: MFCD00049603
Formula:
SeTl₂ MW: 487,73 g/mol Melting Pt: ∼340 °C Density: 9,05 Storage Temperature: Ambient |
MDL Number:
MFCD00049603 CAS Number: 15572-25-5 EINECS: 239-627-1 UN: 1707 ADR: 6.1,II Merck Index: 14,9267 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...