Arsenic(III) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
41708.06
41708.14
ALFA41708.06EA
270
SGD
ALFA41708.06
ALFA41708.14
Arsenic(III) telluride ≥99.999% (metals basis)
Arsenic (III) telluride
Purity: 99.999%
CAS No.: 12044-54-1
Molecular Formula: As2Te3
Formula Weight: 532.64
Physical Form: -160 Mesh Powder
Density: 6.50
Melting Point: 381°
UN No.: 1557
MDL No.: MFCD00014167
Formula:
As₂Te₃ Melting Pt: 381 °C Density: 6,50 Storage Temperature: Ambient |
MDL Number:
MFCD00014167 CAS Number: 12044-54-1 EINECS: 234-955-1 UN: 1557 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...