Copper(I) telluride ≥99.5% (metals basis)

Supplier: Thermo Scientific
Warning

13140.06 13140.14
ALFA13140.06EA 152 SGD
ALFA13140.06 ALFA13140.14
Copper(I) telluride ≥99.5% (metals basis)
Copper (I) telluride

Purity: 99.5% CAS No.: 12019-52-2 Molecular Formula: Cu2Te Formula Weight: 254.68 Physical Form: 3mm & down Pieces Density: 7.27 Refractive Index: 1.45 Appearance: Blue-black Odour: Odourless MDL No.: MFCD00049727

Formula: Cu₂Te
MW: 254,69 g/mol
Melting Pt: 1125 °C
Density: 7,27
Storage Temperature: Ambient
MDL Number: MFCD00049727
CAS Number: 12019-52-2
EINECS: 234-646-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR