Rhenium trioxide ≥99.9% (metals basis)
Supplier: Thermo Scientific
Synonyms:
Rhenium(VI) oxide, Rhenium (VI) oxide
62109.03
62109.06
62109.14
ALFA62109.03EA
332
SGD
ALFA62109.03
ALFA62109.06
ALFA62109.14
Rhenium trioxide ≥99.9% (metals basis)
Rhenium trioxide
Purity: 99.9%
CAS No.: 1314-28-9
Molecular Formula: ReO3
Formula Weight: 236.20
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Violet
Boiling Point: 400° dec.
Merck Reference: 14,8180
MDL No.: MFCD00016306
Formula:
O₃Re MW: 234,21 g/mol Boiling Pt: 400 °C (dec.) Density: 7,4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016306 CAS Number: 1314-28-9 EINECS: 215-228-8 Merck Index: 14,8180 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...