2,7-Dimethoxynaphthalene ≥98%
Supplier: Thermo Scientific
H27971.14
H27971.06
ALFAH27971.14EA
0
SGD
ALFAH27971.14
ALFAH27971.06
2,7-Dimethoxynaphthalene ≥98%
2,7-Dimethoxynaphthalene
Purity: 98%
CAS No.: 3469-26-9
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: 137-140°
MDL No.: MFCD00004064
Formula:
C₁₂H₁₂O₂ MW: 188,23 g/mol Boiling Pt: 299 °C (1013 hPa) Melting Pt: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004064 CAS Number: 3469-26-9 EINECS: 222-433-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...