3-Bromo-2-chloropyridine 97

Supplier: Thermo Fisher Scientific
Danger

B25206.03 B25206.06 B25206.14
ALFAB25206.03EA 0 SGD
ALFAB25206.03 ALFAB25206.06 ALFAB25206.14
3-Bromo-2-chloropyridine 97
3-Bromo-2-chloropyridine

Purity: 97% CAS No.: 52200-48-3 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Melting Point: 52-57° Boiling Point: 97°/10mm MDL No.: MFCD00234007

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 97 °C (10 mmHg)
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00234007
CAS Number: 52200-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR