2,4-Dichloropyridine 97%
Supplier: Thermo Scientific
|
Warning
|
L20040.06
L20040.03
ALFAL20040.06EA
0
SGD
ALFAL20040.06
ALFAL20040.03
2,4-Dichloropyridine 97%
2,4-Dichloropyridine
Purity: 97%
CAS No.: 26452-80-2
Molecular Formula: C5H3Cl2N
Formula Weight: 147.99
Flash Point: 89°(192°F)
Refractive Index: 1.5485
Melting Point: -1°
Boiling Point: 189-190°
Beilstein Reference: 108666
UN No.: 2810
MDL No.: MFCD00955618
Formula:
C₅H₃Cl₂N MW: 147,99 g/mol Boiling Pt: 189…190 °C Melting Pt: –1 °C Density: 1,37 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00955618 CAS Number: 26452-80-2 EINECS: 247-717-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...