2-Chloro-3-methoxypyridine ≥97%

Supplier: Thermo Scientific
Warning

L17696.03 L17696.06
ALFAL17696.03EA 0 SGD
ALFAL17696.03 ALFAL17696.06
2-Chloro-3-methoxypyridine ≥97%
2-Chloro-3-methoxypyridine

Purity: 97% CAS No.: 52605-96-6 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Melting Point: 46-48° Beilstein Reference: 115568

Formula: C₆H₆ClNO
MW: 143,57 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03426022
CAS Number: 52605-96-6
EINECS: 258-039-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR