3-Ethoxy-2-nitropyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
A14371.03
A14371.06
ALFAA14371.03EA
0
SGD
ALFAA14371.03
ALFAA14371.06
3-Ethoxy-2-nitropyridine ≥98%
3-Ethoxy-2-nitropyridine
Purity: 98%
CAS No.: 74037-50-6
Molecular Formula: C7H8N2O3
Formula Weight: 168.15
Flash Point:
Formula:
C₇H₈N₂O₃ MW: 168,15 g/mol Boiling Pt: 156…158 °C (15 mmHg) Melting Pt: 28…32 °C Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006257 CAS Number: 74037-50-6 EINECS: 277-676-0 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...