4-Benzylpyridine ≥99%

Supplier: Thermo Scientific
Danger

A14326.18 A14326.22
ALFAA14326.18EA 59.4 SGD
ALFAA14326.18 ALFAA14326.22
4-Benzylpyridine ≥99%
4-Benzylpyridine

Purity: 99% CAS No.: 2116-65-6 Molecular Formula: C12H11N Formula Weight: 169.23 Flash Point: 115°(239°F) Refractive Index: 1.5805 Melting Point: 9-11° Boiling Point: 286-287° Beilstein Reference: 115831 MDL No.: MFCD00006443

Formula: C₁₂H₁₁N
MW: 169,23 g/mol
Boiling Pt: 286…287 °C
Melting Pt: 9…11 °C
Density: 1,061
Flash Pt: 115 °C (239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006443
CAS Number: 2116-65-6
EINECS: 218-319-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR