2,3,5-Trichlorothiophene ≥97%

Supplier: Thermo Scientific
Danger

L17207.03 L17207.06 L17207.14
ALFAL17207.03EA 0 SGD
ALFAL17207.03 ALFAL17207.06 ALFAL17207.14
2,3,5-Trichlorothiophene ≥97%
2,3,5-Trichlorothiophene

Purity: 97% CAS No.: 17249-77-3 Molecular Formula: C4HCl3S Formula Weight: 187.48 Refractive Index: 1.5790 Melting Point: -16° Boiling Point: 110°/20mm Beilstein Reference: 2431 UN No.: 2810 MDL No.: MFCD01318176

Formula: C₄HCl₃S
MW: 187,48 g/mol
Boiling Pt: 110 °C (20 mmHg)
Melting Pt: –16 °C
Density: 1,586
Storage Temperature: Ambient
MDL Number: MFCD01318176
CAS Number: 17249-77-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR