1-Benzyl-2-pyrrolidinone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Benzylpyrrolidin-2-one

B23538.14 B23538.22
ALFAB23538.14EA 0 SGD
ALFAB23538.14 ALFAB23538.22
1-Benzyl-2-pyrrolidinone ≥97%
1-Benzyl-2-pyrrolidinone

Purity: 97% CAS No.: 5291-77-0 Molecular Formula: C11H13NO Formula Weight: 175.23 Flash Point: 110°(230°F) Refractive Index: 1.5520 Boiling Point: 76-78°/0.1mm Beilstein Reference: 132168 MDL No.: MFCD00003195

Formula: C₁₁H₁₃NO
MW: 175,23 g/mol
Boiling Pt: 76…78 °C (0,1 mmHg)
Density: 1,095
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003195
CAS Number: 5291-77-0
EINECS: 226-131-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR