o-Phenylenediacetonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1,2-Phenylenediacetonitrile, 1,2-Bis(cyanomethyl)-benzene
A13142.06
A13142.14
A13142.18
ALFAA13142.06EA
0
SGD
ALFAA13142.06
ALFAA13142.14
ALFAA13142.18
o-Phenylenediacetonitrile ≥98%
o-Phenylenediacetonitrile
Purity: 98%
CAS No.: 613-73-0
Molecular Formula: C10H8N2
Formula Weight: 156.19
Melting Point: 57-61°
Beilstein Reference: 510075
UN No.: 3439
MDL No.: MFCD00001905
Formula:
C₁₀H₈N₂ MW: 156,19 g/mol Melting Pt: 57…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001905 CAS Number: 613-73-0 EINECS: 210-351-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...