Methyl butyrylacetate ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3-oxohexanoate

L09826.14 L09826.22
ALFAL09826.14EA 79 SGD
ALFAL09826.14 ALFAL09826.22
Methyl butyrylacetate ≥98%
Methyl butyrylacetate

Purity: 98+% CAS No.: 30414-54-1 Molecular Formula: C7H12O3 Formula Weight: 144.17 Flash Point: 90°(194°F) Refractive Index: 1.4260 Melting Point: -43° Boiling Point: 85-90°/5mm Beilstein Reference: 636165 MDL No.: MFCD00040995

Formula: C₇H₁₂O₃
MW: 144,17 g/mol
Boiling Pt: 85…90 °C (5 mmHg)
Melting Pt: –43 °C
Density: 1,037
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040995
CAS Number: 30414-54-1
EINECS: 250-185-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR