2,5-Dichloro-p-benzoquinone ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2,5-Dichloro-1,4-benzoquinone, 2,5-Dichlorobenzo-1,4-quinone

H27152.09 H27152.03
ALFAH27152.09EA 0 SGD
ALFAH27152.09 ALFAH27152.03
2,5-Dichloro-p-benzoquinone ≥98%
2,5-Dichloro-p-benzoquinone

Purity: 98% CAS No.: 615-93-0 Molecular Formula: C6H2Cl2O2 Formula Weight: 176.98 Melting Point: 158-162° MDL No.: MFCD00041736

Formula: C₆H₂Cl₂O₂
MW: 176,99 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00041736
CAS Number: 615-93-0
EINECS: 210-453-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR