Ethanol denatured ≥90% for spectrophotometry (denatured with 5% isopropanol and 5% methanol)
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethyl alcohol denatured
Packaged under argon in resealable ChemSeal™ bottles.
44014.AK
44014.K2
ALFA44014.AKEA
83.7
SGD
ALFA44014.AK
ALFA44014.K2
Ethanol denatured ≥90% for spectrophotometry (denatured with 5% isopropanol and 5% methanol)
Ethanol denatured
Purity: 90%
CAS No.: 64-17-5
Molecular Formula: C2H6O
Formula Weight: 46.08
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.789
Refractive Index: 1.3610
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Alcohol-like
Melting Point: -90°
Boiling Point: 78°
Beilstein Reference: 1718733
Merck Reference: 14,3760
UN No.: 1987
MDL No.: MFCD00003568
Formula:
H₃CCH₂OH MW: 46,07 g/mol Boiling Pt: 78 °C Melting Pt: –90 °C Density: 0,789 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1987 ADR: 3,II Merck Index: 14,3760 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...