Tetrafluorohydroquinone 96%

Supplier: Thermo Scientific
Warning

Synonyms: 2,3,5,6-Tetrafluorohydroquinone, Tetrafluorobenzene-1,4-diol

A17088.03 A17088.06
ALFAA17088.03EA 0 SGD
ALFAA17088.03 ALFAA17088.06
Tetrafluorohydroquinone 96%
Tetrafluorohydroquinone

Purity: 98% CAS No.: 771-63-1 Molecular Formula: C6H2F4O2 Formula Weight: 182.08 Melting Point: 168-170° MDL No.: MFCD00002340

Formula: C₆H₂F₄O₂
MW: 182,07 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002340
CAS Number: 771-63-1
EINECS: 212-237-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR