Water with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK & JACKSON

NP463-20 463-4
BJAANP463-20EA 3090 SGD
BJAANP463-20 BJAA463-4
Water with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0,05% (v/v) formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 100 °C
Density: 0,9983 g/cm³ (20 °C)
Storage Temperature: Ambient
EINECS: 200-579-1
REACH: 01-2119491174-37-XXXX

Order Now

Specification Test Results

Formic Acid (v/v) 0.045 - 0.055%
UV Cutoff <190 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 215 nm < 0.020 AU
HPLC Gradient @ 254 nm < 0.010 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR