4-(4-Chlorophenyl)piperidin-4-ol ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(4-Chlorophenyl)-4-hydroxypiperidine

A12137.03 A12137.06
ALFAA12137.03EA 0 SGD
ALFAA12137.03 ALFAA12137.06
4-(4-Chlorophenyl)piperidin-4-ol ≥97%
4-(4-Chlorophenyl)piperidin-4-ol

Purity: 97% CAS No.: 39512-49-7 Molecular Formula: C11H14ClNO Formula Weight: 211.69 Melting Point: 137-141° Beilstein Reference: 165923

Formula: C₁₁H₁₄ClNO
MW: 211,69 g/mol
Melting Pt: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00006001
CAS Number: 39512-49-7
EINECS: 254-479-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR