3-Cyanocoumarin ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Coumarin-3-carbonitrile, 2-Oxo-2H-chromene-3-carbonitrile
H27871.06
H27871.03
ALFAH27871.06EA
0
SGD
ALFAH27871.06
ALFAH27871.03
3-Cyanocoumarin ≥97%
3-Cyanocoumarin
Purity: 97%
CAS No.: 15119-34-3
Molecular Formula: C10H5NO2
Formula Weight: 171.16
Melting Point: 184-188°
MDL No.: MFCD00115699
Formula:
C₁₀H₅NO₂ MW: 171,16 g/mol Melting Pt: 184…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00115699 CAS Number: 15119-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...