2-Amino-4-(trifluoromethyl)pyrimidine 99%

Supplier: Thermo Scientific
Warning

H50358.03 H50358.06
ALFAH50358.03EA 189 SGD
ALFAH50358.03 ALFAH50358.06
2-Amino-4-(trifluoromethyl)pyrimidine 99%
2-Amino-4-(trifluoromethyl)pyrimidine

Purity: 99% CAS No.: 16075-42-6 Molecular Formula: C5H4F3N3 Formula Weight: 163.10 Melting Point: 68-70° MDL No.: MFCD00828624

Formula: C₅H₄F₃N₃
MW: 163,1 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00828624
CAS Number: 16075-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR