1,2-Phenylenedioxydiacetic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: O-Phenylenedioxydiacetic Acid, Catechol-O,O-diacetic acid, 2,2-(1,2-Phenylenebis(oxy))diacetic acid

L01843.09 L01843.18
ALFAL01843.09EA 105 SGD
ALFAL01843.09 ALFAL01843.18
1,2-Phenylenedioxydiacetic acid ≥97%
1,2-Phenylenedioxydiacetic acid

Purity: 97% CAS No.: 5411-14-3 Molecular Formula: C10H10O6 Formula Weight: 226.18 Melting Point: 177-181° Beilstein Reference: 2057228

Formula: C₁₀H₁₀O₆
MW: 226,19 g/mol
Melting Pt: 177…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00014353
CAS Number: 5411-14-3
EINECS: 226-488-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR