3,5-Diphenyl-1H-pyrazole ≥98%
Supplier: Thermo Scientific
L04904.06
L04904.14
ALFAL04904.06EA
153
SGD
ALFAL04904.06
ALFAL04904.14
3,5-Diphenyl-1H-pyrazole ≥98%
3,5-Diphenyl-1H-pyrazole
Purity: 98+%
CAS No.: 1145-01-3
Molecular Formula: C15H12N2
Formula Weight: 220.28
Melting Point: 199-204°
Beilstein Reference: 158264
Formula:
C₁₅H₁₂N₂ MW: 220,27 g/mol Melting Pt: 199…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039675 CAS Number: 1145-01-3 EINECS: 214-543-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...