2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Amino-5-trifluoromethyl-1,3,4-thiadiazole
A17061.03
A17061.06
A17061.14
ALFAA17061.03EA
0
SGD
ALFAA17061.03
ALFAA17061.06
ALFAA17061.14
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole 98%
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Purity: 98%
CAS No.: 10444-89-0
Molecular Formula: C3H2F3N3S
Formula Weight: 169.13
Melting Point: 222-227°
Beilstein Reference: 513140
Formula:
C₃H₂F₃N₃S MW: 169,13 g/mol Melting Pt: 222…227 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003109 CAS Number: 10444-89-0 EINECS: 233-930-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...