4-Phenyl-1,2,3-thiadiazole
Supplier: Thermo Scientific
|
Warning
|
L20197.03
L20197.06
L20197.MD
ALFAL20197.03EA
0
SGD
ALFAL20197.03
ALFAL20197.06
ALFAL20197.MD
4-Phenyl-1,2,3-thiadiazole
4-Phenyl-1,2,3-thiadiazole
CAS No.: 25445-77-6
Molecular Formula: C8H6N2S
Formula Weight: 162.21
Melting Point: 76-80°
Boiling Point: ca 100°/1.5mm
Beilstein Reference: 119480
Formula:
C₈H₆N₂S MW: 162,22 g/mol Boiling Pt: ∼100 °C (1,5 mmHg) Melting Pt: 76…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00087232 CAS Number: 25445-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...