Diethyl isopropylidenemalonate ≥97%
Supplier: Thermo Scientific
L11195.06
L11195.14
ALFAL11195.06EA
0
SGD
ALFAL11195.06
ALFAL11195.14
Diethyl isopropylidenemalonate ≥97%
Diethyl isopropylidenemalonate
Purity: 97%
CAS No.: 6802-75-1
Molecular Formula: C10H16O4
Formula Weight: 200.24
Flash Point: 109°(228°F)
Refractive Index: 1.4500
Boiling Point: 123-126°/12mm
Beilstein Reference: 1776122
MDL No.: MFCD00009147
Formula:
C₁₀H₁₆O₄ MW: 200,23 g/mol Boiling Pt: 123…126 °C (12 mmHg) Density: 1,021 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009147 CAS Number: 6802-75-1 EINECS: 229-876-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...