3-(Trimethylsilyl)-2-oxazolidinone
Supplier: Thermo Scientific
Warning
|
Synonyms:
3-(Trimethylsilyl)oxazolidin-2-one
41209.03
41209.06
41209.14
ALFA41209.03EA
0
SGD
ALFA41209.03
ALFA41209.06
ALFA41209.14
3-(Trimethylsilyl)-2-oxazolidinone
3-(Trimethylsilyl)-2-oxazolidinone
CAS No.: 43112-38-5
Molecular Formula: C6H13NO2Si
Formula Weight: 159.26
Flash Point: 92°(198°F)
Physical Form: Liquid
Density: 1.013
Refractive Index: 1.4540
Sensitivity: Moisture Sensitive
Boiling Point: 99-100°/6mm
MDL No.: MFCD00010197
Formula:
C₆H₁₃NO₂Si MW: 159,26 g/mol Boiling Pt: 99…100 °C (6 mmHg) Density: 1,013 Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010197 CAS Number: 43112-38-5 EINECS: 256-098-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...