5-Methoxy-1-tetralone ≥97%
Supplier: Thermo Scientific
Synonyms:
5-Methoxy-3,4-dihydronaphthalenone
L15129.03
L15129.06
ALFAL15129.03EA
70.6
SGD
ALFAL15129.03
ALFAL15129.06
5-Methoxy-1-tetralone ≥97%
5-Methoxy-1-tetralone
Purity: 97%
CAS No.: 33892-75-0
Molecular Formula: C11H12O2
Formula Weight: 176.22
Melting Point: 86-91°
Boiling Point: 160-162°/7mm
MDL No.: MFCD00001692
Formula:
C₁₁H₁₂O₂ MW: 176,22 g/mol Boiling Pt: 160…162 °C (7 mmHg) Melting Pt: 86…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001692 CAS Number: 33892-75-0 EINECS: 251-723-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...