7-Methoxy-2-tetralone ≥98%
Supplier: Thermo Scientific
Synonyms:
7-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-7-methoxynaphthalen-2(1H)-one
B22432.03
B22432.06
ALFAB22432.03EA
0
SGD
ALFAB22432.03
ALFAB22432.06
7-Methoxy-2-tetralone ≥98%
7-Methoxy-2-tetralone
Purity: 98%
CAS No.: 4133-34-0
Molecular Formula: C11H12O2
Formula Weight: 176.22
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₁H₁₂O₂ MW: 176,22 g/mol Boiling Pt: 124…126 °C (1,5 mmHg) Density: 1,130 Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00001730 CAS Number: 4133-34-0 EINECS: 223-954-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...