4-Biphenylacetonitrile ≥97%
Supplier: Thermo Scientific
|
Warning
|
L19243.04
L19243.ME
ALFAL19243.04EA
153
SGD
ALFAL19243.04
ALFAL19243.ME
4-Biphenylacetonitrile ≥97%
4-Biphenylacetonitrile
Purity: 97%
CAS No.: 31603-77-7
Molecular Formula: C14H11N
Formula Weight: 193.25
Melting Point: 93-97°
Boiling Point: 146-150°/0.03mm
MDL No.: MFCD00016403
Formula:
C₁₄H₁₁N MW: 193,25 g/mol Boiling Pt: 146…150 °C (0,03 mmHg) Melting Pt: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016403 CAS Number: 31603-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...