4-Fluoro-3-methoxyphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (4-Fluoro-3-methoxyphenyl)boronic acid, 4-Fluoro-3-methoxybenzeneboronic acid

H52635.MD H52635.03
ALFAH52635.MDEA 0 SGD
ALFAH52635.MD ALFAH52635.03
4-Fluoro-3-methoxyphenylboronic acid ≥98%
4-Fluoro-3-methoxyphenylboronic acid

Purity: 98% CAS No.: 854778-31-7 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Melting Point: 189-191° MDL No.: MFCD08056358

Formula: C₇H₈BFO₃
MW: 169,95 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD08056358
CAS Number: 854778-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR