3-(3-Fluoro-4-methylphenyl)propionic acid ≥97%

Supplier: Thermo Scientific
Warning

H32105.MD H32105.03
ALFAH32105.MDEA 0 SGD
ALFAH32105.MD ALFAH32105.03
3-(3-Fluoro-4-methylphenyl)propionic acid ≥97%
3-(3-Fluoro-4-methylphenyl)propionic acid

Purity: 97% CAS No.: 881189-62-4 Molecular Formula: C10H11FO2 Formula Weight: 182.19 Melting Point: 95-97° MDL No.: MFCD09025364

Formula: C₁₀H₁₁FO₂
MW: 182,19 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09025364
CAS Number: 881189-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR