2-Nitropyridine ≥97%
Supplier: Thermo Scientific
|
Danger
|
H33657.03
H33657.06
ALFAH33657.03EA
0
SGD
ALFAH33657.03
ALFAH33657.06
2-Nitropyridine ≥97%
2-Nitropyridine
Purity: 97%
CAS No.: 15009-91-3
Molecular Formula: C5H4N2O2
Formula Weight: 124.10
Melting Point: 69-73°
UN No.: 2811
MDL No.: MFCD00160414
Formula:
C₅H₄N₂O₂ MW: 124,10 g/mol Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160414 CAS Number: 15009-91-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...