3,5-Xylylacetonitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 3,5-Dimethylphenylacetonitrile

L10170.09 L10170.04
ALFAL10170.09EA 533 SGD
ALFAL10170.09 ALFAL10170.04
3,5-Xylylacetonitrile ≥98%
3,5-Xylylacetonitrile

Purity: 98% CAS No.: 39101-54-7 Molecular Formula: C10H11N Formula Weight: 145.20 Melting Point: 42-45° Boiling Point: 170°/20mm UN No.: 3439 MDL No.: MFCD00060304

Formula: C₁₀H₁₁N
MW: 145,20 g/mol
Boiling Pt: 170 °C (20 mmHg)
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00060304
CAS Number: 39101-54-7
EINECS: 254-292-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR