Fluorene-4-carboxylic acid 96%

Supplier: Thermo Scientific
Warning

L01164.03 L01164.06 L01164.MD
ALFAL01164.03EA 0 SGD
ALFAL01164.03 ALFAL01164.06 ALFAL01164.MD
Fluorene-4-carboxylic acid 96%
Fluorene-4-carboxylic acid

Purity: 96% CAS No.: 6954-55-8 Molecular Formula: C14H10O2 Formula Weight: 210.23 Melting Point: 191-192° Beilstein Reference: 2050637

Formula: C₁₄H₁₀O₂
MW: 210,23 g/mol
Melting Pt: 191…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00001112
CAS Number: 6954-55-8
EINECS: 230-138-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR