5-Bromo-2-methoxyphenylboronic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromo-2-methoxybenzeneboronic acid

H52497.03 H52497.06
ALFAH52497.03EA 0 SGD
ALFAH52497.03 ALFAH52497.06
5-Bromo-2-methoxyphenylboronic acid ≥97%
5-Bromo-2-methoxyphenylboronic acid

Purity: 97% CAS No.: 89694-45-1 Molecular Formula: C7H8BBrO3 Formula Weight: 230.85 Melting Point: 129-131° MDL No.: MFCD00093409

Formula: C₇H₈BBrO₃
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00093409
CAS Number: 89694-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR