3-Chloro-5-methoxyphenylboronic acid ≥94%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Chloro-5-methoxybenzeneboronic acid

H27442.MD H27442.03
ALFAH27442.MDEA 0 SGD
ALFAH27442.MD ALFAH27442.03
3-Chloro-5-methoxyphenylboronic acid ≥94%
3-Chloro-5-methoxyphenylboronic acid

Purity: 94% CAS No.: 915201-07-9 Molecular Formula: C7H8BClO3 Formula Weight: 186.40 Melting Point: 185-189° MDL No.: MFCD03095046

Formula: C₇H₈BClO₃
MW: 186,40 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03095046
CAS Number: 915201-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR