4-Chloro-2-methoxyphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-2-methoxybenzeneboronic acid, (4-Chloro-2-methoxyphenyl)boronic acid

H52529.MD H52529.03
ALFAH52529.MDEA 0 SGD
ALFAH52529.MD ALFAH52529.03
4-Chloro-2-methoxyphenylboronic acid ≥98%
4-Chloro-2-methoxyphenylboronic acid

Purity: 98% CAS No.: 762287-57-0 Molecular Formula: C7H8BClO3 Formula Weight: 186.40 Melting Point: 128-134° MDL No.: MFCD02684317

Formula: C₇H₈BClO₃
MW: 186,40 g/mol
Melting Pt: 128…134 °C
Storage Temperature: Ambient
MDL Number: MFCD02684317
CAS Number: 762287-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR