2-Nitro-4-(trifluoromethyl)phenylacetonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
L19239.03
L19239.06
ALFAL19239.03EA
0
SGD
ALFAL19239.03
ALFAL19239.06
2-Nitro-4-(trifluoromethyl)phenylacetonitrile ≥98%
2-Nitro-4-(trifluoromethyl)phenylacetonitrile
Purity: 98%
CAS No.: 13544-06-4
Molecular Formula: C9H5F3N2O2
Formula Weight: 230.15
Odour: Characteristic
Melting Point: 38-42°
Boiling Point: 134-138°/0.5mm
Beilstein Reference: 2588629
UN No.: 3439
MDL No.: MFCD00045621
Formula:
C₉H₅F₃N₂O₂ MW: 230,15 g/mol Boiling Pt: 134…138 °C (0,5 mmHg) Melting Pt: 38…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045621 CAS Number: 13544-06-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...