5-Fluoro-2-methoxybenzyl bromide ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-(Bromomethyl)-4-fluoro-1-methoxybenzene

H25957.14 H25957.03 H25957.06
ALFAH25957.14EA 0 SGD
ALFAH25957.14 ALFAH25957.03 ALFAH25957.06
5-Fluoro-2-methoxybenzyl bromide ≥98%
5-Fluoro-2-methoxybenzyl bromide

Purity: 98+% CAS No.: 700381-18-6 Molecular Formula: C8H8BrFO Formula Weight: 219.05 Melting Point: 58-62° UN No.: 3261 MDL No.: MFCD00671769

Formula: C₈H₈BrFO
MW: 219,05 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00671769
CAS Number: 700381-18-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR