(±)-3-Methyl-3-heptanol ≥98%

Supplier: Thermo Scientific
Warning

B20959.03 B20959.06 B20959.14
ALFAB20959.03EA 0 SGD
ALFAB20959.03 ALFAB20959.06 ALFAB20959.14
(±)-3-Methyl-3-heptanol ≥98%
(±)-3-Methyl-3-heptanol

Purity: 98% CAS No.: 5582-82-1 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 58°(136°F) Boiling Point: 64°/201mm UN No.: 1987 MDL No.: MFCD00021841

Formula: C₈H₁₈O
MW: 130,23 g/mol
Boiling Pt: 64 °C (201 mmHg)
Density: 0,83
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021841
CAS Number: 5582-82-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR