5-Bromo-2-(trifluoromethoxy)toluene ≥97%

Supplier: Thermo Scientific
Warning

H32119.03 H32119.06
ALFAH32119.03EA 0 SGD
ALFAH32119.03 ALFAH32119.06
5-Bromo-2-(trifluoromethoxy)toluene ≥97%
5-Bromo-2-(trifluoromethoxy)toluene

Purity: 97% CAS No.: 887268-26-0 Molecular Formula: C8H6BrF3O Formula Weight: 255.04 Physical Form: Liquid Appearance: Colourless to organge MDL No.: MFCD04972695

Formula: C₈H₆BrF₃O
MW: 255,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972695
CAS Number: 887268-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR