(Pentafluoroethyl)trimethylsilane ≥97%

Supplier: Thermo Scientific
Danger

H25775.03 H25775.06 H25775.14
ALFAH25775.03EA 0 SGD
ALFAH25775.03 ALFAH25775.06 ALFAH25775.14
(Pentafluoroethyl)trimethylsilane ≥97%
(Pentafluoroethyl)trimethylsilane

Purity: 97% CAS No.: 124898-13-1 Molecular Formula: C5H9F5Si Formula Weight: 192.21 Refractive Index: 1.3270

Formula: C₅H₉F₅Si
MW: 192,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116436
CAS Number: 124898-13-1
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR