(3,3,3-Trifluoropropyl)dichloromethylsilane ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Dichloro(methyl)(3,3,3-trifluoropropyl)silane, (3,3,3-Trifluoropropyl)methyldichlorosilane
L16669.06
L16669.14
L16669.22
ALFAL16669.06EA
0
SGD
ALFAL16669.06
ALFAL16669.14
ALFAL16669.22
(3,3,3-Trifluoropropyl)dichloromethylsilane ≥97%
(3,3,3-Trifluoropropyl)dichloromethylsilane
Purity: 97%
CAS No.: 675-62-7
Molecular Formula: C4H7Cl2F3Si
Formula Weight: 211.09
Flash Point: 15°(59°F)
Refractive Index: 1.3850
Boiling Point: 121-122°
Beilstein Reference: 1744964
UN No.: 2985
MDL No.: MFCD00039294
Formula:
C₄H₇Cl₂F₃Si MW: 211,09 g/mol Boiling Pt: 121…122 °C Density: 1,261 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039294 CAS Number: 675-62-7 EINECS: 211-623-4 UN: 2985 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...