2,2'-Dichloro-4,4'-bipyridyl 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2,2'-Dichloro-4,4'-bipyridine

H27695.MC H27695.ME
ALFAH27695.MCEA 0 SGD
ALFAH27695.MC ALFAH27695.ME
2,2'-Dichloro-4,4'-bipyridyl 95%
2,2'-Dichloro-4,4'-bipyridyl

Purity: 95% CAS No.: 53344-74-4 Molecular Formula: C10H6Cl2N2 Formula Weight: 225.08 Melting Point: 240-243° MDL No.: MFCD01106345

Formula: C₁₀H₆Cl₂N₂
MW: 225,08 g/mol
Melting Pt: 240…243 °C
Storage Temperature: Ambient
MDL Number: MFCD01106345
CAS Number: 53344-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR