Methyl hydrogen glutarate ≥95%

Supplier: Thermo Scientific

Synonyms: 5-Methoxy-5-oxopentanoic acid

L09196.09 L09196.18
ALFAL09196.09EA 134 SGD
ALFAL09196.09 ALFAL09196.18
Methyl hydrogen glutarate ≥95%
Methyl hydrogen glutarate

Purity: 95% CAS No.: 1501-27-5 Molecular Formula: C6H10O4 Formula Weight: 146.14 Flash Point: 110°(230°F) Refractive Index: 1.4380 Boiling Point: 150-152°/12mm Beilstein Reference: 1762381 MDL No.: MFCD00004409

Formula: C₆H₁₀O₄
MW: 146,1412 g/mol
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00004409
CAS Number: 1501-27-5
EINECS: 216-116-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR