Creatine (N-amidinosarcosine), anhydrous 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine
A17477.14
A17477.22
ALFAA17477.14EA
0
SGD
ALFAA17477.14
ALFAA17477.22
Creatine (N-amidinosarcosine), anhydrous 98%
Creatine (N-Amidinosarcosine)
Purity: 98%
CAS No.: 57-00-1
Molecular Formula: C4H9N3O2
Formula Weight: 131.13
Physical Form: Powder
Density: 1.33
Appearance: White
Melting Point: ca 290° dec.
Beilstein Reference: 907175
Merck Reference: 14,2568
MDL No.: MFCD00004282
Formula:
C₄H₉N₃O₂ MW: 131,13 g/mol Melting Pt: ∼ 290 °C (dec.) Density: 1,33 Storage Temperature: Ambient |
MDL Number:
MFCD00004282 CAS Number: 57-00-1 EINECS: 200-306-6 Merck Index: 14,2568 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...