4-Bromo-2-fluoro-6-nitrophenol 98

Supplier: Thermo Fisher Scientific
Danger

H32153.03 H32153.06
ALFAH32153.03EA 0 SGD
ALFAH32153.03 ALFAH32153.06
4-Bromo-2-fluoro-6-nitrophenol 98
4-Bromo-2-fluoro-6-nitrophenol

Purity: 98% CAS No.: 320-76-3 Molecular Formula: C6H3BrFNO3 Formula Weight: 236.00 MDL No.: MFCD00013372

Formula: C₆H₃BrFNO₃
MW: 236.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013372
CAS Number: 320-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR