5-Quinolineboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: Quinolin-5-ylboronic acid

L19639.03 L19639.MD
ALFAL19639.03EA 0 SGD
ALFAL19639.03 ALFAL19639.MD
5-Quinolineboronic acid ≥97%
5-Quinolineboronic acid

Purity: 97% CAS No.: 355386-94-6 Molecular Formula: C9H8BNO2 Formula Weight: 172.98 Melting Point: 144-148° MDL No.: MFCD03095058

Formula: C₉H₈BNO₂
MW: 172,98 g/mol
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD03095058
CAS Number: 355386-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR