2'-Deoxyguanosine hydrate ≥99%
Supplier: Thermo Scientific
Synonyms:
9-(2-Deoxy-β-D-ribofuranosyl)guanine hydrate, Guanine-2′-deoxyriboside hydrate, 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one hydrate(1:x)
Purity: 99%
CAS No.: 961-07-9
Molecular Formula: C10H13N5O4.xH2O
Formula Weight: 267.25(anhy)
Beilstein Reference: 39814
Optical Rotation: -32° (c=0.5 in water)
Formula:
C₁₀H₁₃N₅O₄·xH₂O MW: 267,24 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150760 CAS Number: 961-07-9 EINECS: 213-505-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...